Skip to Content
Merck
All Photos(1)

Key Documents

456306

Sigma-Aldrich

Benz[a]anthracene-d12

98 atom % D

Synonym(s):

1,2-Benzanthracene-d12, Tetraphene-d12

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€399.00

€399.00


Check Cart for Availability

Request more information

Select a Size

Change View
250 MG
€399.00

About This Item

Empirical Formula (Hill Notation):
C18D12
CAS Number:
Molecular Weight:
240.36
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.12

€399.00


Check Cart for Availability

Request more information

isotopic purity

98 atom % D

Quality Level

Assay

99% (CP)

form

solid

mp

157-159 °C (lit.)

application(s)

electronics

mass shift

M+12

SMILES string

[2H]c1c([2H])c([2H])c2c(c1[2H])c([2H])c([2H])c3c([2H])c4c([2H])c([2H])c([2H])c([2H])c4c([2H])c23

InChI

1S/C18H12/c1-2-7-15-12-18-16(11-14(15)6-1)10-9-13-5-3-4-8-17(13)18/h1-12H/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D,11D,12D

InChI key

DXBHBZVCASKNBY-AQZSQYOVSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yusuke Yamamoto et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1000, 187-191 (2015-08-08)
Polycyclic aromatic hydrocarbons (PAHs) are formed from the incomplete combustion or pyrolysis of organic matter during industrial processing and various human activities, but human exposure to PAHs has not yet been elucidated in detail. To assess long-term exposure to PAHs

Articles

Compare PAH determination in olive oil: traditional GB 5009.265-2021 method vs. faster, simpler SPE with Supelclean EZ-POP NP; both meet criteria.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service