Skip to Content
Merck
All Photos(1)

Key Documents

364614

Sigma-Aldrich

Chrysene-d12

98 atom % D

Synonym(s):

Dodecadeuteriochrysene

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€71.90
50 MG
€113.00
250 MG
€347.00

€71.90


Check Cart for Availability

Request more information

Select a Size

Change View
25 MG
€71.90
50 MG
€113.00
250 MG
€347.00

About This Item

Empirical Formula (Hill Notation):
C18D12
CAS Number:
Molecular Weight:
240.36
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.12

€71.90


Check Cart for Availability

Request more information

isotopic purity

98 atom % D

Quality Level

Assay

99% (CP)

bp

448 °C (lit.)

mp

252-254 °C (lit.)

mass shift

M+12

SMILES string

[2H]c1c([2H])c([2H])c2c(c1[2H])c([2H])c([2H])c3c4c([2H])c([2H])c([2H])c([2H])c4c([2H])c([2H])c23

InChI

1S/C18H12/c1-3-7-15-13(5-1)9-11-18-16-8-4-2-6-14(16)10-12-17(15)18/h1-12H/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D,11D,12D

InChI key

WDECIBYCCFPHNR-AQZSQYOVSA-N

Looking for similar products? Visit Product Comparison Guide

Application

May be used as an analytical standard

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaowei Wang et al.
Journal of separation science, 37(14), 1842-1849 (2014-04-29)
An automatic exposed and in-syringe dynamic single-drop microextraction method (SDME) for the determination of five polycyclic musks in natural waters was developed using gas chromatography with mass spectrometry. Online agitation was first introduced to the automatic SDME with a magnetic
Zheng-Xia Chen et al.
Chemosphere, 131, 192-200 (2015-04-12)
Fine roots of woody plants are a heterogeneous system differing markedly in structure and function. Nevertheless, knowledge about the plant uptake of organic pollutants via fine roots is scarce to date. In the present study, plant uptake, translocation, and return
Célia A Alves et al.
Environmental science and pollution research international, 22(15), 11526-11542 (2015-04-02)
The main purpose of this work was to evaluate the chemical composition of particulate matter (PM) emitted by eight different light-duty vehicles. Exhaust samples from petrol and diesel cars (Euro 3 to Euro 5) were collected in a chassis dynamometer
Dennis S Mkhize et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 79(5), 808-819 (2019-04-27)
A passive sampling device, based on molecularly imprinted membranes (MIM), was fabricated and optimised for sampling polychlorinated biphenyls (PCBs) in aquatic ecosystems. The newly-developed passive sampler was subjected to in-situ calibration studies to determine PCB sampling rates under various conditions
Chen-Chou Wu et al.
Environmental science & technology, 49(17), 10607-10615 (2015-08-11)
Barbecuing or charcoal-grilling has become part of popular outdoor recreational activities nowadays; however, potential human health hazards through outdoor exposure to barbecue fumes have yet to be adequately quantified. To fill this knowledge gap, atmospheric size-fractioned particle and gaseous samples

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service