Skip to Content
Merck
All Photos(1)

Documents

616664

Sigma-Aldrich

Benzo[e]pyrene-d12

98 atom % D, 98% (CP)

Synonym(s):

4,5-Benzopyrene-d12

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20D12
CAS Number:
Molecular Weight:
264.38
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % D

Quality Level

Assay

98% (CP)

mp

177-180 °C (lit.)

mass shift

M+12

SMILES string

[2H]c1c([2H])c([2H])c2c(c1[2H])c3c([2H])c([2H])c([2H])c4c([2H])c([2H])c5c([2H])c([2H])c([2H])c2c5c34

InChI

1S/C20H12/c1-2-8-16-15(7-1)17-9-3-5-13-11-12-14-6-4-10-18(16)20(14)19(13)17/h1-12H/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D,11D,12D

InChI key

TXVHTIQJNYSSKO-AQZSQYOVSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Marco Schintu et al.
Marine pollution bulletin, 135, 819-827 (2018-10-12)
Passive sampling techniques were used for monitoring trace metals and polycyclic aromatic hydrocarbons (PAHs) in the seawater surrounding the Costa Concordia shipwreck (Isola del Giglio, Italy). The monitoring lasted two and a half years (2012-2014) and considered all four phases
Badreddine Barhoumi et al.
Marine environmental research, 101, 184-195 (2014-08-12)
In this study, biological responses and contaminant levels in biological tissues were investigated in grass goby fish specimens (Zosterisessor ophiocephalus) collected from five stations in a moderately polluted ecosystem, namely the Bizerte lagoon on the north coast of Tunisia. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service