![Benz[a]anthracene-d12 98 atom % D](/deepweb/assets/sigmaaldrich/product/structures/119/003/f3ccbf30-1771-4e29-b7dc-ad1e33659269/640/f3ccbf30-1771-4e29-b7dc-ad1e33659269.png)
Benz[a]anthracene-d12
![Benz[a]anthracene-d12 98 atom % D](/deepweb/assets/sigmaaldrich/product/structures/119/003/f3ccbf30-1771-4e29-b7dc-ad1e33659269/640/f3ccbf30-1771-4e29-b7dc-ad1e33659269.png)
Synonym(s):
1,2-Benzanthracene-d12, Tetraphene-d12
Empirical Formula (Hill Notation):
C18D12
CAS Number:
Molecular Weight:
240.36
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Isotope
Application
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing