Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
material
stainless steel
packaging
pkg of 1 ea
manufacturer/tradename
VICI Valco® ZT1C
bore size
0.25 mm
fitting
10-32 female Valco for 1/16 in. O.D. tubing (fittings included)
compatibility
for connecting 1/16 in.
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description zero dead volume, for connecting 1/16 in., bore size 0.25 mm | description zero dead volume, for connecting 1/16 in., bore size 0.75 mm | description for connecting 1/16 in., bore size 0.25 mm | description internal reducing, for connecting 1/16 to 1/32 in., bore size 0.25 mm |
| material stainless steel | material - | material stainless steel union and fittings | material stainless steel union and fittings |
| manufacturer/tradename VICI Valco® ZT1C | manufacturer/tradename VICI Valco® ZT1 | manufacturer/tradename VICI Valco® ZU1C | manufacturer/tradename VICI Valco® ZRU1.5 |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
| fitting 10-32 female Valco for 1/16 in. O.D. tubing (fittings included) | fitting - | fitting - | fitting 10-32 Valco for 1/16 in. O.D. tubing (nuts and ferrules included), 6-32 Valco for 1/32 in. O.D. tubing (nuts and ferrules included) |
| compatibility for connecting 1/16 in. | compatibility for connecting 1/16 in. | compatibility for connecting 1/16 in. | compatibility for connecting 1/16 to 1/32 in. |
| bore size 0.25 mm | bore size 0.75 mm | bore size 0.25 mm | bore size 0.25 mm |
Still not finding the right product?
Explore all of our products under T-Stück
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?