58627
Synonym(e):
Valco® Verbinder
Eigenschaften
stainless steel union and fittings
internal
pkg of 1 ea
VICI Valco® ZU1C
0.25 mm
for connecting 1/16 in.
Beschreibung
Unterschiede anzeigen
1 of 1
Supelco
Verbinder
Z227242
Kopplung
57659
22949
Verbinder, reduzierend
for connecting 1/16 in., bore size 0.25 mm
PEEK, for connecting 1/16 in., bore size 0.25 mm
bore 0.020 in., 10-32 thread
zero dead volume, for connecting 1/8 to 1/16 in., bore size 0.25 mm
female
-
PEEK
stainless steel
VICI Valco® IZR21C
for connecting 1/8 to 1/16 in.
Explore all of our products under Verbinder
Hier finden Sie alle aktuellen Versionen:
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumente section.