Fortfahren mit
23804
Kapillarsäulenstoßverbinder
body only (ferrules not included)
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Größe/SKUVerfügbarkeitPreis
Über diesen Artikel
UNSPSC Code:
41115700
NACRES:
SB.54
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
packaging
pkg of 1 ea
technique(s)
gas chromatography (GC): suitable
General description
This device consists of a double-tapered ferrule and a stainless steel compression body with a threaded nut. Small (2.3 cm x 0.6 cm) and light (4.4 g with ferrule), it provides a gas tight seal without a change in column efficiency or inertness. The columns to be connected can have the same or different internal and external diameters. The butt connection is made inside the special double-tapered ferrule. The ferrule is then compressed within the housing. When the column ends are butted squarely and tightly together, the butt connector will not alter the chromatographic performance of your capillary columns. There is little or no dead volume and little chance of gas flow disruption by following these steps:
  • Make sure the bore of the ferrule is clean. Blow out any ferrule fragments with nitrogen. Using a magnifier, examine the column ends to be connected. Make sure each cut is clean and square. The two ends must butt squarely, without any gaps.
  • With white typewriter correction fluid, place a reference mark 1/4 inch from the end of the column with the larger bore. This mark will help you to confirm visually that the end of the column is centered within the 1/2 inch ferrule.
  • Place the ferrule inside the housing and loosely tighten the nut. Feed the unmarked column completely through the ferrule and out the opposite end. Cut off ~1 inch (25 mm) of the column to ensure no ferrule fragments are in the column. Draw the column back far enough to insert the marked column into the ferrule to the indicating mark. Tighten the nut about 1/8 turn past fingertight.
  • Press the ends of the columns together, observing the reference mark to make certain they butt together at the center of the ferrule. Tighten the ferrule to about 1/4-1/2 turn past fingertight. Gently pull on both columns to ensure they are secure. If they are loose, additional tightening is necessary.
Any undetected leaking connection, including this butt connection, can allow oxygen and water vapor to enter the system. Leak check the butt connector in the same manner as any capillary column connection. DO NOT USE LIQUID LEAK INDICATORS. Liquids can contaminate the capillary system. We recommend using a GOW-MAC electronic leak detector. These thermal conductivity detectors are highly sensitive to trace amounts of hydrogen or helium, and will not contaminate the system.
Capillary column butt connector is made by inserting two capillaries into a piece of PTFE shrinking tubing, which when heat is applied secures the two capillaries in place. It has shown to have no contribution to band spreading.
Other Notes
We offer a variety of chromatography accessories including analytical syringes
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
237962047923627
description
body only (ferrules not included)
description
I.D. 0.4 mm,
Supeltex M-2 ferrule included
description
pkg of 12 ea
description
pkg of 5 ea
packaging
pkg of 1 ea
packaging
pkg of 1 ea
packaging
pkg of 12 ea
packaging
pkg of 5 ea
technique(s)
gas chromatography (GC): suitable
technique(s)
gas chromatography (GC): suitable
technique(s)
gas chromatography (GC): suitable
technique(s)
gas chromatography (GC): suitable
Still not finding the right product?
Explore all of our products under Kapillarsäulenstoßverbinder
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)