Ansicht ändern
| Größe/SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
UNSPSC Code:
41115700
NACRES:
SB.54
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltenpackaging
pkg of 5 ea
technique(s)
gas chromatography (GC): suitable
General description
GlasSeal connectors quickly connect fused silica tubing of the same or different diameter with no tools and no leaks. Use them to connect a guard column or transfer line, repair a broken column, or connect two columns (same or different phases).
A Y connector splits a sample to two columns or a column effluent to two detectors. All of these connectors are silanized for an inert inside surface. Choose borosilicate glass or fused silica. Use with our 0.25mm-0.53mm I.D. tubing.
A Y connector splits a sample to two columns or a column effluent to two detectors. All of these connectors are silanized for an inert inside surface. Choose borosilicate glass or fused silica. Use with our 0.25mm-0.53mm I.D. tubing.
Other Notes
We offer a variety of chromatography accessories including analytical syringes
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description pkg of 5 ea | description pkg of 25 ea | description pack of 20 ea | description I.D. 0.4 mm, Supeltex M-2 ferrule included |
| packaging pkg of 5 ea | packaging pkg of 25 ea | packaging pack of 20 ea | packaging pkg of 1 ea |
| technique(s) gas chromatography (GC): suitable | technique(s) gas chromatography (GC): suitable | technique(s) - | technique(s) gas chromatography (GC): suitable |
Still not finding the right product?
Explore all of our products under GlasSeal Kapillarsäulenverbinder, Quarzglas
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?