Fortfahren mit
22970
Adustable Single Port Mini-Vap Evaporator/Concentrator
Mini-Vap size 1 1/2 in. × 8 1/2 in. , adjustable,
single unit
Synonym(e):
Mini-Vap 1mv ea
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Größe/SKUVerfügbarkeitPreis
Über diesen Artikel
UNSPSC Code:
41122001
NACRES:
NB.43
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
agency
suitable for EPA OTM-45
packaging
pkg of 1 ea
technique(s)
chromatography: suitable
Mini-Vap size
1 1/2 in. × 8 1/2 in. , adjustable
application(s)
PFAS testing
General description
The Mini-Vap includes a needle valve for fine metering of air or nitrogen drying gas. Ideal for a wide variety of needs in evaporation or concentration.
The adjustable Mini-Vap (22970) is a versatile concentrator/evaporator that can be used with a single container. The unique design allows for any size container, from the smallest vial to a 250 mL beaker.
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
22971EP5305000363RAB1773
description
Mini-Vap size 1 1/2 in. × 8 1/2 in. , adjustable,
single unit
description
Mini-Vap L × W 7 1/2 in. (19 cm) × 1 1/2 in. (4 cm), for use with 1-250mL containers, pkg of 1 ea
description
complete system, integrated vacuum pump, with rotor F-45-48-11, AC/DC input 230 V (GB plug)
description
for serum, plasma and cell culture supernatants
application(s)
PFAS testing
application(s)
PFAS testing
application(s)
-
application(s)
-
agency
suitable for EPA OTM-45
agency
suitable for EPA 1633, suitable for EPA 533, suitable for GB 31604.35-2016, suitable for EPA 537.1, suitable for DIN 38407-42, suitable for EPA OTM-45, suitable for ISO/CEN 15968-2010, suitable for ISO 25101
agency
-
agency
-
Mini-Vap size
1 1/2 in. × 8 1/2 in. , adjustable
Mini-Vap size
-
Mini-Vap size
-
Mini-Vap size
-
packaging
pkg of 1 ea
packaging
pkg of 1 ea
packaging
1 ea of
packaging
-
EPAOTM45
HJ1334-2023, FDAC-010_03, EPAmethod533, GB31604_35-2016, DIN17681-2, CEN-TS15968-2010, DIN38414-14, DIN38407-42, DIN17681-1, FDAC-010_02, EPA1633, ISO21675_2019, HJ1333-2023, DIN17892, EPA537_1
vacuum concentrator for gentle sample concentration, high-efficiency vacuum centrifuge for nucleotides, Eppendorf Concentrator Plus troubleshooting guide, Eppendorf Concentrator Plus user manual, chemical-resistant vacuum concentrator for acids, fast concentration vacuum centrifuge for labs, Eppendorf Concentrator Plus ergonomic design, Eppendorf Concentrator Plus parts and accessories, Eppendorf Concentrator Plus with coated lid, laboratory-grade vacuum concentrator for proteins, Eppendorf Concentrator Plus specifications and features, Eppendorf vacuum concentrator for DNA and RNA, Eppendorf Concentrator Plus vacuum centrifuge
-
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)