Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
packaging
pkg of 100 ea
technique(s)
gas chromatography (GC): suitable using :Suitable
I.D.
1/8 in.
fitting
1/8 in. for 1/8 in. O.D. packed column
compatibility
1/8 in. O.D. Column
General description
Material: keramikgefülltes PTFE
Höchsttemperatur: 250 °C
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description I.D. 1/8 in., 1/8 in. O.D. Column, pkg of 100 ea | description I.D. 1/8 in., 1/8 in. O.D. Column, pkg of 50 ea | description I.D. 1/4 in., 1/4 in. O.D. Column, pkg of 10 ea | description I.D. 1/4 in., 1/4 in. O.D. Column, pkg of 50 ea |
| I.D. 1/8 in. | I.D. 1/8 in. | I.D. 1/4 in. | I.D. 1/4 in. |
| technique(s) gas chromatography (GC): suitable using :Suitable | technique(s) - | technique(s) gas chromatography (GC): suitable using :Suitable | technique(s) gas chromatography (GC): suitable using :Suitable |
| packaging pkg of 100 ea | packaging pkg of 50 ea | packaging pkg of 10 ea | packaging pkg of 50 ea |
| fitting 1/8 in. for 1/8 in. O.D. packed column | fitting - | fitting - | fitting 1/4 in. GC fitting ferrule for 1/4 in. O.D. packed column |
| compatibility 1/8 in. O.D. Column | compatibility 1/8 in. O.D. Column | compatibility 1/4 in. O.D. Column | compatibility 1/4 in. O.D. Column |
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?