Fortfahren mit
20977
Hamilton® GASTIGHT® Spritze, Serie 1700, Luer-Spitze
1702LT, volume 25 μL, without needle, Luer Tip
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Größe/SKUVerfügbarkeitPreis
Über diesen Artikel
UNSPSC Code:
41122001
NACRES:
NB.03
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
glass syringe
needle size
26 ga
feature
without needle Luer Tip
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 80201
volume
25 μL
application(s)
general analytical
General description
Hamilton syringe is available in gas-tight type that minimizes the gas exchange in GC analysis. It gives accurate volume delivery. The plunger is mostly unsurpassed. The tip is threaded in a luer lock syringe allowing the needle to be tightly screwed to the tip.
Application
It was used to inject nutritional supplement the study animals (rats & mice) to understand choroidal neovascularization development and reduction of acute conjunctivitis inflammation.
Legal Information
GASTIGHT is a registered trademark of Hamilton Co.
Hamilton is a registered trademark of Hamilton Co.
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
207172620320714
description
1702LT, volume 25 μL, without needle, Luer Tip
description
1725LT, volume 250 μL, without needle, Luer Tip, needle size (not included)
description
1710TLL, volume 100 μL, PTFE Luer lock, needle size (not included)
description
1750LT, volume 0.5 mL, without needle, Luer Tip, needle size (not included)
material
glass syringe
material
glass body
material
PTFE Luer lock
material
glass body
packaging
pkg of 1 ea
packaging
pkg of 1 ea
packaging
pkg of 1 ea
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 80201
manufacturer/tradename
Hamilton 81101
manufacturer/tradename
Hamilton 81020
manufacturer/tradename
Hamilton 81201
volume
25 μL
volume
250 μL
volume
100 μL
volume
0.5 mL
feature
without needle Luer Tip
feature
without needle Luer Tip
feature
-
feature
without needle Luer Tip
needle size
26 ga
needle size
(not included)
needle size
(not included)
needle size
(not included)
Still not finding the right product?
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)