Z744250
Wilmad® 50 ml Suprasil® Small Volume Liquid Nitrogen Dewar
for Bruker®, O.D. 8 mm, pack of 1 ea
Synonym(e):
EPR cryo dewar, EPR dewar
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.15
Empfohlene Produkte
Verpackung
pack of 1 ea
Hersteller/Markenname
Wilmad WG-816-D-Q
AD
8 mm
Allgemeine Beschreibung
Dewar Flasks provide a convenient alternative to Variable Temperature systems for studies at 77 K, and are silvered around the reservoir. 50 mL flasks last 15 minutes after a filling.
Rechtliche Hinweise
Bruker is a registered trademark of Bruker-Physik AG
Suprasil is a registered trademark of HERAEUS QUARZGLAS GMBH & CO.
Wilmad is a registered trademark of SP Industries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)