Z737712
Jenco™ compound microscopes
AC/DC input 220 V AC, brightfield, achromatic lens, binocular, mechanical stage; led or other illumination
Synonym(e):
compound microscopes, microscopes
Alle Fotos(1)
About This Item
UNSPSC-Code:
41161502
NACRES:
MA.44
Empfohlene Produkte
Beschreibung
3-plug
EU 2-prong plug
Hersteller/Markenname
Jenco CP135-B4-2
Allgemeine Beschreibung
- Illumination can be a mirror, a rechargeable LED, an incandescent bulb or halogen bulb
- Eye pieces come in monocular heads, or teaching heads for assisting students in finding specimens and also binocular heads for easy viewing with both eyes
- Objectives are offered in multiple quality and quantity levels
- Video systems are available to project images on a screen
- Industry leading 5 year warranty
Science teachers cannot expend their entire budget on microscopes or sacrifice quality on such an integral tool, so Jenco keeps prices low, quality high and offers selection to provide them affordable options. Jenco brand microscopes virtually eliminate downtime and our optics facilitate student throughput.
Choose from different frames for the right reason and teach science with the right equipment for your students.
Choose from different frames for the right reason and teach science with the right equipment for your students.
Rechtliche Hinweise
Jenco is a trademark of Jenco International, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)