Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|---|---|
960 ea | Warenkorb auf Verfügbarkeit prüfen | € 85,60 |
4800 ea | Warenkorb auf Verfügbarkeit prüfen | € 311,00 |
Über diesen Artikel
€ 85,60
material
colorless , polypropylene
sterility
sterile
feature
barrier: no, graduations
packaging
case of 4800 ea (50 racks of 96 tips), rack of 96, pack of 960 ea (10 racks of 96 tips)
manufacturer/tradename
Sorenson 31790T
maximum volume
300 μL
volume
300 μL
General description
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description maximum volume 300 μL, rack of 96, sterile | description maximum volume 200 μL, pkg of 96 tips CellRack, sterile | description maximum volume 300 μL, case of 1000 ea (Bag), non-sterile | description maximum volume 300 μL, rack of 96, non-sterile |
| manufacturer/tradename Sorenson 31790T | manufacturer/tradename Sorenson 15280T | manufacturer/tradename Sorenson 31770T | manufacturer/tradename Sorenson 317800T |
| sterility sterile | sterility sterile | sterility non-sterile | sterility non-sterile |
| volume 300 μL | volume 200 μL | volume 300 μL | volume 300 μL |
| feature barrier: no, graduations | feature barrier: no | feature graduations, barrier: no | feature barrier: no, graduations |
| material polypropylene , colorless | material colorless , natural tip, polypropylene | material polypropylene , colorless | material polypropylene , colorless |
| packaging pack of 960 ea (10 racks of 96 tips), rack of 96, case of 4800 ea (50 racks of 96 tips) | packaging pkg of 96 tips CellRack, case of 4800 ea (50 racks of 96 tips), pack of 960 ea (10 racks of 96 tips) | packaging case of 1000 ea (Bag), case of 10000 ea (10 bags of 1000 tips) | packaging case of 4800 ea (50 racks of 96 tips), rack of 96, pack of 960 ea (10 racks of 96 tips) |
Still not finding the right product?
Explore all of our products under Sorenson™ low binding standard tips
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?