Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|---|---|
960 ea | Warenkorb auf Verfügbarkeit prüfen | € 79,30 |
4800 ea | Warenkorb auf Verfügbarkeit prüfen | € 294,00 |
Über diesen Artikel
€ 79,30
material
colorless , polypropylene
sterility
non-sterile
feature
barrier: no, graduations
packaging
case of 4800 ea (50 racks of 96 tips), rack of 96, pack of 960 ea (10 racks of 96 tips)
manufacturer/tradename
Sorenson 317800T
maximum volume
300 μL
volume
300 μL
General description
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description maximum volume 300 μL, rack of 96, non-sterile | description maximum volume 200 μL, pkg of 96 QuickRack, non-sterile | description maximum volume 200 μL, pkg of 96 tips CellRack, non-sterile | description pack of 8 ea (Empty racks), non-sterile |
| manufacturer/tradename Sorenson 317800T | manufacturer/tradename Sorenson 30450T | manufacturer/tradename Sorenson 15270T | manufacturer/tradename Sorenson 28310 |
| volume 300 μL | volume 200 μL | volume 200 μL | volume - |
| feature barrier: no, graduations | feature barrier: no, graduations | feature barrier: no | feature - |
| sterility non-sterile | sterility non-sterile | sterility non-sterile | sterility non-sterile |
| material polypropylene , colorless | material colorless , polypropylene | material polypropylene , colorless | material polypropylene , colorless |
| packaging case of 4800 ea (50 racks of 96 tips), pack of 960 ea (10 racks of 96 tips), rack of 96 | packaging case of 4800 ea (50 racks of 96 tips), pack of 960 ea (10 racks of 96 tips), pkg of 96 QuickRack | packaging pkg of 96 tips CellRack, case of 4800 ea (50 racks of 96 tips), pack of 960 ea (10 racks of 96 tips) | packaging pack of 8 ea (Empty racks) |
Still not finding the right product?
Explore all of our products under Sorenson™ low binding standard tips
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?