Fortfahren mit
Z659630
Bandelin SONOREX™ Ultrasonic bath accessories
insert tub with lid for Z659584, Z659592 and Z659002
Synonym(e):
Bandelin 715, holder, retainer, sonification holder, sonification tub sonifier insert
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Größe/SKUVerfügbarkeitPreis
1 ea
Warenkorb auf Verfügbarkeit prüfen
€ 104,00
Über diesen Artikel
NACRES:
NB.74
UNSPSC Code:
41103706
eCl@ss:
32090120
€ 104,00
Warenkorb auf Verfügbarkeit prüfen
​
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
polyethylene
description
Bandelin KW 3
packaging
pack of 1 ea
manufacturer/tradename
Bandelin 715
availability
not available in USA and Canada
L × W × H
195 mm × 115 mm × 88 mm
General description
Stainless steel lid for an ultrasonic bath protects the bath liquid from external contamination and prevents unpleasant odors from the cleaning or disinfection agents added. In addition, the noise emitted when the ultrasonic cleaner is in operation, which may be perceived as annoying, is reduced. The lids are designed in such a way that any condensation that occurs is directed back into the bath.
Features and Benefits
  • protects the bath fluid from contamination
  • sound-absorbing
Legal Information
SONOREX is a trademark of Bandelin Electronic GmbH & Co. KG
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
Z659738Z659487Z660116
description
insert tub with lid for Z659584, Z659592 and Z659002
description
insert tub with lid for Z659673 and Z659029
description
tank lid to fit Z659460
description
insert basket for Z659037
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
material
polyethylene
material
polyethylene
material
stainless steel
material
stainless steel
availability
not available in USA and Canada
availability
not available in USA and Canada
availability
-
availability
not available in USA and Canada
description
Bandelin KW 3
description
Bandelin KW 5
description
Bandelin D 08
description
Bandelin K 10 B
L × W × H
195 mm × 115 mm × 88 mm
L × W × H
-
L × W × H
-
L × W × H
-
manufacturer/tradename
Bandelin 715
manufacturer/tradename
Bandelin 240
manufacturer/tradename
Bandelin 218
manufacturer/tradename
Bandelin 230
Still not finding the right product?
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)