Merck
TMO342002-9050
Nalgene® sterile diagnostic bottle
high-density polyethylene, bottle capacity 15 mL, natural, case of 896 ea
Synonym(e):
small volume plastic bottle, test kit bottle
Größe auswählen
896 EACH
€ 839,00
€ 839,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
896 EACH
€ 839,00
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.43
€ 839,00
Check Cart for Availability
Materialien
high-density polyethylene
natural
Sterilität
sterile
Verpackung
case of 896 ea
Hersteller/Markenname
Nalgene 342002-9050
Flaschenkapazität
15 mL
Allgemeine Beschreibung
Perform many small volume packing applications with Thermo Scientific Nalgene HDPE Diagnostic Bottles with Closure. Bottles can be used in test kits and are packed in shrink-wrapped SBS trays for easy handling. Trays can be used as an aid for machine fill-line loading. They come assembled with closures without a liner that can wrinkle.
Rechtliche Hinweise
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)