Merck
TMO2401-1000
Nalgene® economy wash bottle
low-density polyethylene, volume 1000 mL, unlabelled, case of 12 ea
Synonym(e):
LDPE flush bottle, LDPE wash bottle
Größe auswählen
12 EACH
€ 122,00
€ 122,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
12 EACH
€ 122,00
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.43
€ 122,00
Check Cart for Availability
Materialien
low-density polyethylene
Verpackung
case of 12 ea
Hersteller/Markenname
Nalgene 2401-1000
Kapazität
1000 mL
Volumen
1000 mL
Allgemeine Beschreibung
Prevent separation when squeezing Thermo Scientific Nalgene LDPE Economy Wash Bottles, featuring a tight-fitting draw tube and closure. These low density polyethylene bottles are designed to dispense through the top with a closure and stem molded in one piece for uniform leakproof† service. Cut back tip to increase flow.
Rechtliche Hinweise
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)