Merck
TMO174888
Nunclon® cell culture dishes
diam. 60 mm, with lid, Nunclon® Delta treated, case of 120 ea
Synonym(e):
cell culture dish, cell culture treateddish
Größe auswählen
500 EACH
€ 1.600,00
€ 1.600,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
500 EACH
€ 1.600,00
About This Item
UNSPSC-Code:
41121812
NACRES:
NB.13
€ 1.600,00
Check Cart for Availability
Materialien
polystyrene dishes with lids
Sterilität
sterile; γ-irradiated
Verpackung
case of 120 ea
Hersteller/Markenname
Nunc 174888
Durchmesser
60 mm
Allgemeine Beschreibung
  • Large range available, treated for cell culture but with many other uses
  • Optically clear and very flat for use in microscope
  • Non-toxic
  • Certified surface treatment for optimal cell attachment and growth
Rechtliche Hinweise
Nunclon is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)