Merck
TMO140668
Nunc® cell culture inserts in 6-deepwell Nunclon Delta surface plate
cell culture treated, polycarbonate, sterile, pore size 8.0 μm, case of 24 ea (supplied in 4 x 6 well plates)
Synonym(e):
multidish membrane insert, treated membrane insert
Größe auswählen
24 EACH
€ 281,00
€ 281,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
24 EACH
€ 281,00
About This Item
UNSPSC-Code:
41122102
NACRES:
NB.13
€ 281,00
Check Cart for Availability
Materialien
polycarbonate
Sterilität
sterile
Verpackung
case of 24 ea (supplied in 4 x 6 well plates)
Hersteller/Markenname
Nunc 140668
Porengröße
8.0 μm pore size
Bindungsart
cell culture treated
Allgemeine Beschreibung
Cultivate most cell types without matrix coating when using Thermo Scientific Nunc Polycarbonate Membrane Inserts in Multidishes.
Rechtliche Hinweise
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)