Fortfahren mit
Spend € 450 — Save 5 %; Spend € 700 — Save 10 %; Spend € 1.500 — Save 15 %*
Anmelden zur Überprüfung der Spielberechtigung. Produkte kaufen
Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|---|---|
24 ea | Warenkorb auf Verfügbarkeit prüfen | € 278,00 |
Über diesen Artikel
€ 278,00
Sparen Sie bis zu 15 %
material
polycarbonate
sterility
sterile
packaging
case of 24 ea (supplied in 4 x 6 well plates)
manufacturer/tradename
Nunc 140660
pore size
0.4 μm pore size
binding type
cell culture treated
General description
Application
- Tumor migration and invasion assays.
- Epithelial (e.g. Caco-2) and endothelial (e.g. brain microvascular endothelial cells) barriers assays.
- Dermal/epidermal and epithelial tissue models (e.g. artificial skin model).
- Chemotaxis studies.
- Co-culture.
Features and Benefits
- Tissue culture-treated porous membrane for optimized cell attachment.
- Three pore sizes: 0.4, 3, and 8 micron.
- Pre-packed in sterile Nunclon® Delta-treated multi-well plates.
- Nontoxic and nonpyrogenic.
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description cell culture treated, polycarbonate, sterile, pore size 0.4 μm, case of 24 ea (supplied in 4 x 6 well plates) | description cell culture treated, polycarbonate, sterile, pore size 8.0 μm, case of 24 ea (supplied in 4 x 6 well plates) | description cell culture treated, polycarbonate, sterile, pore size 0.4 μm, case of 24 ea (supplied in 4 x 6 well plates) | description cell culture treated, polycarbonate, sterile, pore size 3.0 μm, case of 24 ea (supplied in 4 x 6 well plates) |
| sterility sterile | sterility sterile | sterility sterile | sterility sterile |
| binding type cell culture treated | binding type cell culture treated | binding type cell culture treated | binding type cell culture treated |
| manufacturer/tradename Nunc 140660 | manufacturer/tradename Nunc 140668 | manufacturer/tradename Nunc 140640 | manufacturer/tradename Nunc 140663 |
| material polycarbonate | material polycarbonate | material polycarbonate | material polycarbonate |
| packaging case of 24 ea (supplied in 4 x 6 well plates) | packaging case of 24 ea (supplied in 4 x 6 well plates) | packaging case of 24 ea (supplied in 4 x 6 well plates) | packaging case of 24 ea (supplied in 4 x 6 well plates) |
| pore size 0.4 μm pore size | pore size 8.0 μm pore size | pore size 0.4 μm pore size | pore size 3.0 μm pore size |
Still not finding the right product?
Explore all of our products under Nunc® cell culture inserts in 6-deepwell Nunclon Delta surface plate
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?