Fortfahren mit
T4412
Tweezers
Style #5, Fine needle-sharp, non-magnetic alloy (tips may be restraightened when bent)
Synonym(e):
fine forceps, fine tweezers, needle tweezers, Ted Pella 525, laboratory forceps
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Ihnen/SKUVerfügbarkeitPreis
1 ea
Warenkorb auf Verfügbarkeit prüfen
€ 210,00
Über diesen Artikel
NACRES:
MA.01
UNSPSC Code:
27112105
€ 210,00
Warenkorb auf Verfügbarkeit prüfen
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
non-magnetic alloy (tips may be restraightened when bent)
packaging
pack of 1 ea
manufacturer/tradename
Ted Pella 525
size
110 mm
General description
Specifically designed for microscopy applications. All are ultra-high precision tools of Swiss manufacture.
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
T4162T4787T4912
Tweezers Style #5, Fine needle-sharp, non-magnetic alloy (tips may be restraightened when bent)
T4412
Tweezers
Tweezers Style #3C, very sharp, non-magnetic alloy (tips may be restraightened when bent)
T4162
Tweezers
Tweezers Style #7, sharp, hooked, non-magnetic alloy (tips may be restraightened when bent)
T4787
Tweezers
Tweezers Style #7, sharp, hooked, anti-magnetic stainless steel
T4912
Tweezers
description
Style #5, Fine needle-sharp, non-magnetic alloy (tips may be restraightened when bent)
description
Style #3C, very sharp, non-magnetic alloy (tips may be restraightened when bent)
description
Style #7, sharp, hooked, non-magnetic alloy (tips may be restraightened when bent)
description
Style #7, sharp, hooked, anti-magnetic stainless steel
material
non-magnetic alloy (tips may be restraightened when bent)
material
non-magnetic alloy (tips may be restraightened when bent)
material
non-magnetic alloy (tips may be restraightened when bent)
material
anti-magnetic stainless steel
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
manufacturer/tradename
Ted Pella 525
manufacturer/tradename
Ted Pella 523-C
manufacturer/tradename
Ted Pella 527
manufacturer/tradename
Ted Pella 507-NM
size
110 mm
size
110 mm
size
115 mm
size
115 mm
Still not finding the right product?
Explore all of our products under Tweezers
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)