Größe auswählen
| Ihnen/SKU | Verfügbarkeit | Preis |
|---|---|---|
1 ea | Warenkorb auf Verfügbarkeit prüfen | € 112,00 |
Über diesen Artikel
€ 112,00
material
anti-magnetic stainless steel
packaging
pack of 1 ea
manufacturer/tradename
Ted Pella 507-NM
size
115 mm
General description
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description Style #7, sharp, hooked, anti-magnetic stainless steel | description Style #7, sharp, hooked, PTFE-coated, anti-magnetic stainless steel | description Style #3C, very sharp, anti-magnetic stainless steel | description Style #5, Fine needle-sharp, anti-magnetic stainless steel |
| material anti-magnetic stainless steel | material PTFE-coated, anti-magnetic stainless steel | material anti-magnetic stainless steel | material anti-magnetic stainless steel |
| manufacturer/tradename Ted Pella 507-NM | manufacturer/tradename Ted Pella 5627-TEF | manufacturer/tradename Ted Pella 503C-NM | manufacturer/tradename Ted Pella 505-NM |
| size 115 mm | size 115 mm | size 110 mm | size 110 mm |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea |
Still not finding the right product?
Explore all of our products under Tweezers
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?