Merck
MTCR1040Y
MTC Bio Fraction CollectorTube Rack
holds 80 x 1.5 /2.0 mL tubes, yellow, pack of 5 ea
Synonym(e):
sample slide, stacking western blot box, tube holder, tube storage
About This Item
UNSPSC-Code:
41122804
NACRES:
NB.12
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Materialien
polypropylene
yellow
Leistungsmerkmale
holds 80 x 1.5 /2.0 mL tubes
Verpackung
pack of 5 ea
Hersteller/Markenname
MTC Bio R1040-Y
Allgemeine Beschreibung
Compact, convenient 80 well racks designed to keep your tubes neatly and efficiently organized. Theses autoclavable polypropylene racks are compatible with conical and self-standing microtubes and all wells have raised, molded numbers for quick identification. The bottom of each well has been recessed to improve conical tube stability without sacrificing the stability of self-standing tubes.
Tube Rack, fraction collector type, 80 x 1.5mL/2mL, yellow, pack of 5
Leistungsmerkmale und Vorteile
  • 80 well (5 rows of 16) for 1.5 to 2.0mL microtubes
  • Rainbow pack available — 5 colors
  • Two frosted marking areas, on the bottom and side
  • Raised numbers beside each well
  • Recessed well bottoms for improved tube stability
Rechtliche Hinweise
MTC is a trademark of MTC Bio
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)