Größe auswählen
| Ihnen/SKU | Verfügbarkeit | Preis |
|---|---|---|
1 ea | Warenkorb auf Verfügbarkeit prüfen | € 81,00 |
Über diesen Artikel
€ 81,00
material
acrylic , green (dispenser)
feature
holds Small laboratory consumables, items, and accessories
packaging
pack of 1 ea
manufacturer/tradename
Heathrow Scientific HS23411
L × W × H
180 mm × 155 mm × 165 mm
General description
Features and Benefits
- Lid remains open when tilted back
- Non-skid rubber feet and pre-drilled holes for wall mounting
- Dual compartment with removable divider
- Mounting screws included for the option of fixing to a wall or raised surface
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description green (dispenser) | description blue | description clear (bin) | description clear (bin) |
| manufacturer/tradename Heathrow Scientific HS23411 | manufacturer/tradename Heathrow Scientific HS23412 | manufacturer/tradename Heathrow Scientific HS23402 | manufacturer/tradename Heathrow Scientific HS23404 |
| feature holds Small laboratory consumables, items, and accessories | feature holds Small laboratory consumables | feature holds Pipette tips, tubes, finger cots, fittings, and other small bulk items, accessories | feature holds Pipette tips, tubes, finger cots, fittings, and other small bulk items, accessories |
| material acrylic , green (dispenser) | material acrylic , blue | material acrylic , clear (bin) | material acrylic , clear (bin) |
| L × W × H 180 mm × 155 mm × 165 mm | L × W × H 155 mm × 155 mm × 170 mm | L × W × H 150 mm × 177 mm × 368 mm | L × W × H 287 mm × 170 mm × 217 mm |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea |
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?