Ansicht ändern
| Ihnen/SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
NACRES:
NB.22
UNSPSC Code:
41122800
Material:
polypropylene (corrugated)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltenmaterial
polypropylene (corrugated)
packaging
pack of 10 trays
manufacturer/tradename
Heathrow Scientific
L × W × H
16 × 10.24 × 3.5 in.
size
24 wells
color
white
General description
Corrugated storage trays can be used with the included dividers or sample containers or remove them for general lab product storage and transport.
Features and Benefits
- moisture resistant, significantly reducing chance of mold
- lightweight and sturdy to hold up to 24 full sample containers
- stackable with other trays
- removable dividers allow outer shell to be used for storage or transport of laboratory products
- pack includes 10 trays and 10 dividers
- disposable
- supplied flat-packed to reduce storage space
Legal Information
Droplet is a trademark of Heathrow Scientific LLC
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description polypropylene (corrugated), holds 24 sample containers, pack of 10 trays | description pack of 1 tray and 1 insert | description slot size 1 × 3 in., positions, 20, polystyrene, pack of 6 trays | description with inserts and 16 mm test tube rack |
| material polypropylene (corrugated) | material - | material polystyrene | material - |
| packaging pack of 10 trays | packaging pack of 1 tray and 1 insert | packaging pack of 6 trays | packaging - |
| size 24 wells | size - | size - | size - |
| color white | color - | color - | color - |
| L × W × H 16 × 10.24 × 3.5 in. | L × W × H - | L × W × H - | L × W × H - |
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?