Fortfahren mit
HS1042
3-in-1 Dispenser Counter or Wall Mount
gray
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Größe/SKUVerfügbarkeitPreis
1 ea
Warenkorb auf Verfügbarkeit prüfen
€ 199,00
Über diesen Artikel
NACRES:
NB.13
UNSPSC Code:
41121800
Manufacturer/tradename:
Heathrow Scientific HS1042
Feature:
holds safety glasses, earplugs, soft covers (including bouffant caps, beard covers, shoe covers) and small items, accessories
Material:
acrylic , gray
Packaging:
pack of 1 ea
€ 199,00
Warenkorb auf Verfügbarkeit prüfen
​
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
acrylic , gray
feature
holds safety glasses, earplugs, soft covers (including bouffant caps, beard covers, shoe covers) and small items, accessories
packaging
pack of 1 ea
manufacturer/tradename
Heathrow Scientific HS1042
L × W × D
406 mm × 406 mm × 203 mm
General description
The 3-in-1 Dispenser′s separate lift-up lids make loading and dispensing supplies easy. Three-compartment dispenser can be used to organize safety glasses, earplugs, soft covers (including bouffant caps, beard covers, shoe covers) or any small items that need to be kept within reach.
Features and Benefits
  • The unit includes screws for wall mounting or can be set on a counter
  • Middle compartment has reach-in hole for dispensing
  • Durable acrylic is see-through so product levels are always visible
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
HS1040CHS1040BHS1040A
description
gray
description
clear
description
green (dispenser)
description
-
feature
holds safety glasses, earplugs, soft covers (including bouffant caps, beard covers, shoe covers) and small items, accessories
feature
holds Soft covers (including bouffant caps, beard covers, shoe covers) and small items, accessories
feature
holds Small laboratory consumables, items, and accessories
feature
-
manufacturer/tradename
Heathrow Scientific HS1042
manufacturer/tradename
Heathrow Scientific HS1040C
manufacturer/tradename
Heathrow Scientific HS1040B
manufacturer/tradename
Heathrow Scientific HS1040A
material
acrylic , gray
material
acrylic , clear
material
green (dispenser), acrylic
material
blue acrylic
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
-
L × W × D
406 mm × 406 mm × 203 mm
L × W × D
-
L × W × D
-
L × W × D
-
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)