Merck
EP0030124332
Eppendorf® PCR tubes
colorless, capacity 0.2 mL, pack of 1000 ea
Synonym(e):
eppendorf tubes, microcentrifuge tubes, microfuge tubes
Größe auswählen
1 PKG
€ 165,00
€ 165,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 PKG
€ 165,00
About This Item
UNSPSC-Code:
41121800
NACRES:
NC.21
€ 165,00
Check Cart for Availability
Materialien
cap (plug seal)
colorless
colorless cap
colorless tube
conical bottom
Leistungsmerkmale
graduations: no
neck thread Push fit
Verpackung
pack of 1000 ea
Hersteller/Markenname
Eppendorf® 0030124332
Kapazität
0.2 mL
Leistungsmerkmale und Vorteile
  • The hinged lid with a defined position and the tight sealing prevents contamination and evaporation.
  • The lid is etched to ensure labeling is convenient.
  • Designed to have maximum transparency even on the tube base
  • Easy to open
  • Suitable with all thermal cyclers with 0.2 mL block format
  • The product is batch-tested and certified free of DNA, DNase, RNase and PCR inhibitors (PCR clean).
Rechtliche Hinweise
Eppendorf is a registered trademark of Eppendorf AG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)