Merck
EP0030106300
Eppendorf® UVette®, disposable single seated cUVette®s
pkg of 80 ea
Synonym(e):
Eppendorf UVette
Größe auswählen
80 EA
€ 69,60
€ 69,60
Voraussichtliches Versanddatum02. April 2025
Größe auswählen
Ansicht ändern
80 EA
€ 69,60
About This Item
UNSPSC-Code:
41121813
NACRES:
NB.44
€ 69,60
Voraussichtliches Versanddatum02. April 2025
Verpackung
pkg of 80 ea
Hersteller/Markenname
Eppendorf® 0030106300
Leistungsmerkmale und Vorteile
  • Eppendorf® UVette® disposable single seated cuvettes are perfect for applications outside of the UV range, such as colorimetric protein assays (Bradford, Lowry, etc.), determining of the optical density of bacterial cultures (OD600 methods), and kinetic and fluorescence measurements.
  • Individually packaged.
  • PCR-clean and protein-free quality.
  • UV transparent plastic, free of fluoropolymers and other halogenated hydrocarbons.
  • Two built-in optical path lengths in a single cuvette - just turn the UVette 90° to change from 10 mm and 2 mm.
  • Volume markings for 500 μL and 1000 μL.
  • Individually packaged and certified PCR clean and protein-free quality for sensitive (e.g. RNA) and valuable samples.
  • Deep-lying optical window prevents scratches.
  • Tapered cuvette base for optimal filling, frosted grip for labeling.
  • Ideal for use in the Eppendorf BioPhotometer and Eppendorf BioSpectrometer, adapters available for use with other common spectrophotometers.
  • Routine pack for convenient access to each UVette and safe storage in a reclosable box.
Rechtliche Hinweise
Eppendorf is a registered trademark of Eppendorf AG
UVette is a registered trademark of Eppendorf AG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)