Merck
BR759115
BRAND® semi-micro cuvette
PMMA, minimum filling volume 1.5 mL
Synonym(e):
Cuvettes
Größe auswählen
100 EA
€ 15,40
€ 15,40
Check Cart for Availability
Größe auswählen
Ansicht ändern
100 EA
€ 15,40
About This Item
UNSPSC-Code:
41121813
NACRES:
SB.32
€ 15,40
Check Cart for Availability
Materialien
PMMA
Verpackung
pack of 100 ea
Hersteller/Markenname
BRAND 759115
Allgemeine Beschreibung
  • BRAND® semi-micro cuvette is manufactured under controlled room conditions.
  • Clear, clean optical path with an indication of optical path orientation.
  • Packed grouped by mold cavity number to ensure the lowest variation of extinction coefficient.
  • Standard 10mm light path.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)