Größe auswählen
| Ihnen/SKU | Verfügbarkeit | Preis |
|---|---|---|
5 ea | Warenkorb auf Verfügbarkeit prüfen | € 326,00 |
Über diesen Artikel
€ 326,00
material
cap (fill)
sterility
sterile
packaging
case of 5
manufacturer/tradename
Corning 3283
I.D.
9.5 mm
size
7.94 mm , barbed fitting
General description
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description I.D. 9.5 mm, size 7.94 mm , barbed fitting, sterile, cap (fill), case of 5 | description sterile, male MPC coupling, with male end cap, 6.4 mm I.D. barbed fitting with female end cap, cap (fill), case of 4 | description sterile, male MPC coupling with male end cap, 9.5 mm I.D. barbed fitting with female end cap, cap (fill), case of 4 | description sterile, 6.4 mm I.D. tubing, 70 cm length, male MPC coupling with female end cap, cap (fill), case of 4 |
| sterility sterile | sterility sterile | sterility sterile | sterility sterile |
| I.D. 9.5 mm | I.D. - | I.D. - | I.D. - |
| manufacturer/tradename Corning 3283 | manufacturer/tradename Corning 3334 | manufacturer/tradename Corning 3339 | manufacturer/tradename Corning 3333 |
| material cap (fill) | material cap (fill) | material cap (fill) | material cap (fill) |
| packaging case of 5 | packaging case of 4 | packaging case of 4 | packaging case of 4 |
| size 7.94 mm , barbed fitting | size - | size - | size - |
Still not finding the right product?
Explore all of our products under Corning® CellSTACK® Zubehör
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?