Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
packaging
pkg of 1 unit
compatibility
for use with Milli-Q® IX 7003, for use with Milli-Q® IX 7005, for use with Milli-Q® IX 7010, for use with Milli-Q® IX 7015, for use with Milli-Q® IQ 7003, for use with Milli-Q® IQ 7005, for use with Milli-Q® IQ 7010, for use with Milli-Q® IQ 7015
General description
Diese Halterung montiert Milli-Q® IQ 7003/05/10/15 und Milli-Q® IX 7003/05/10/15 Lagertanks.
Application
Features and Benefits
- Einfach zu montierendes Zubehör.
- Spart Platz auf dem Labortisch.
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description Installation accessory for 25 or 50 L water storage unit, For use with Milli-Q® IQ and Milli-Q® IX systems | description Installation accessory for use with Milli-Q® IQ, Milli-Q® IX and Milli-Q® EQ water purification systems | description For installation of Milli-Q Advantage/Integral/Reference/Direct, Simplicity, Synergy and Elix Reference systems | description For installation of RiOs™ 3, RiOs-DI® 3 and Direct-Q® 3 systems |
| compatibility for use with Milli-Q® IQ 7005, for use with Milli-Q® IX 7015, for use with Milli-Q® IX 7010, for use with Milli-Q® IX 7003, for use with Milli-Q® IQ 7003, for use with Milli-Q® IQ 7010, for use with Milli-Q® IX 7005, for use with Milli-Q® IQ 7015 | compatibility for use with Milli-Q® IX 7015, for use with Milli-Q® IQ 7005, for use with Milli-Q® EQ 7016, for use with Milli-Q® IQ 7010, for use with Milli-Q® IX 7005, for use with Milli-Q® IQ 7000, for use with Milli-Q® IQ 7015, for use with Milli-Q® IX 7010, for use with Milli-Q® IQ 7003, for use with Milli-Q® IX 7003, for use with Milli-Q® EQ 7008 | compatibility for use with Milli-Q® Reference, for use with Simplicity® UV, for use with Elix® Advantage, for use with Milli-Q® Advantage A10, for use with Synergy® UV, for use with Elix® Reference, for use with Milli-Q® Integral, for use with Milli-Q® Direct | compatibility for use with RiOs-DI® UV, for use with Direct-Q®, for use with RiOs-DI® Clinical, for use with RiOs™, for use with Direct-Q® UV |
| packaging pkg of 1 unit | packaging pkg of 1 unit | packaging pkg of 1 unit | packaging pkg of 1 unit |
Still not finding the right product?
Explore all of our products under Wandmontagehalterung
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?