Merck
Z742594
Accuris SmartDoc UV blocking mat
For blocking UV light
Größe auswählen
1 EA
€ 53,40
€ 53,40
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
€ 53,40
About This Item
UNSPSC-Code:
41106000
NACRES:
NB.79
€ 53,40
Check Cart for Availability
Leistungsmerkmale
For blocking UV light
Verpackung
pkg of 1
Hersteller/Markenname
Benchmark Scientific E5000-MAT
Allgemeine Beschreibung
Accuris SmartDoc mat for blocking UV light for use with UV illuminators with viewing surface greater than 18 x 18 cm.
Rechtliche Hinweise
Accuris is a trademark of Benchmark Scientific, Inc.
SmartDoc is a trademark of Benchmark Scientific, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Questions
  1. what colour is it? dose it transparent?
    1 answer
    1. The color of the UV Blocking mat is black, as shown in the image on the product detail page. This product is not transparent.