Merck
Z684864
Quick-release clamps
without extension, for 150 mm (6 in.) Schott flange
Synonym(e):
clamps, quick release clamp
Größe auswählen
1 EA
€ 238,00
€ 238,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
€ 238,00
About This Item
UNSPSC-Code:
41122413
NACRES:
NB.43
€ 238,00
Check Cart for Availability
Materialien
stainless steel
Sterilität
non-sterile
Verpackung
case of 1
Hersteller/Markenname
Ace Glass 6517-27
Allgemeine Beschreibung
SS clamps for use with reaction flasks and heads with a 100 mm Schott flange. The extension rod can be clamped to the support frame and helps stabilize the reaction assembly.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)