Merck
Z153966
Ausgießringe für Duran® Laborflaschen
GL 45 neck thread, red melamine resin
Synonym(e):
DURAN pouring ring, ETFE, GL45, high temperature, pouring ring
Größe auswählen
10 EA
€ 17,60
€ 17,60
Check Cart for Availability
Größe auswählen
Ansicht ändern
10 EA
€ 17,60
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.17
€ 17,60
Check Cart for Availability
Materialien
red melamine resin
Hersteller/Markenname
DURAN®
Schott
Parameter
-100-180 °C temp. stability
Fitting
GL 45 neck thread
Allgemeine Beschreibung
The red polybutylene terephthalate (PBT) GL cap and ETFE pouring rings offer greater thermal and chemical resistance than the equivalent polypropylene components. Screw cap seal has a PTFE faced cap liner for efficient sealing. ETFE pouring ring ensures drip free pouring.
Leistungsmerkmale und Vorteile
Ausgußringe verhindern Tropfenbildung beim Ausgießen von Duran Flaschen
Rechtliche Hinweise
DURAN is a registered trademark of DWK Life Sciences
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Questions
  1. Hello how many times can Shott bottle plastic lids and poring rings be autoclaved at 121 degrees, and what is the best practices for replacement ?
    1 answer
    1. The reusability of this product has not been tested. However, the material is polypropylene, which can be used multiple times. It is difficult to provide an expected service life for DURAN® screw closures. Application variables, including cleaning conditions, sterilization processes, and media types in contact with the articles, all affect the likely service life.