Merck
Z680575
Ausgießringe für Duran® Laborflaschen
PTFE, neck thread GL 80
Synonym(e):
pouring ring
Größe auswählen
5 EA
€ 97,20
€ 97,20
Check Cart for Availability
Größe auswählen
Ansicht ändern
5 EA
€ 97,20
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.17
€ 97,20
Check Cart for Availability
Materialien
PTFE
Hersteller/Markenname
DURAN®
Schott
Allgemeine Beschreibung
The material used for this GLS 80 closure is a special compound based on polyarylsulphone. As a result, the chemical, thermal and mechanical properties of the material are improved compared to polypropylene. Thanks to the shortened triple thread, the DURAN® GLS 80® bottle can be opened or closed with only a three-quarter turn. The seal, which is coated on both sides with PTFE, ensures the bottle can be tightly sealed (cap liner: platinum-cured silicone). A matching PTFE pouring ring is also available, permitting clean, drip-free use.
Leistungsmerkmale und Vorteile
Ausgußringe verhindern Tropfenbildung beim Ausgießen von Duran Flaschen
Rechtliche Hinweise
DURAN is a registered trademark of DWK Life Sciences
GLS 80 is a registered trademark of Duran Group
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)