Merck
Y906549
Sigma-Aldrich
Valve
brass diaphragm, for CGA 350 right angle path, external, left hand thread
About This Item
UNSPSC-Code:
12142200
NACRES:
NA.12
Note: This product is only available with the purchase of a bulk gas product and not for individual sale. For more information, please contact Stable Isotopes Customer Service
Materialien
brass diaphragm
Qualitätsniveau
Parameter
2640 psi max. pressure
Länge
4.62 in.
Gewicht
633 g
Fitting
for CGA 350 right angle path, external, left hand thread
Verwandte Kategorien
Sonstige Hinweise
This product is only available with the purchase of a bulk gas product and not for individual sale. For more information, please contact Stable Isotopes Customer Service
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)