Merck
SYNM431004
Synthware all-glass vacuum/inert gas manifold with hollow high vacuum stopcocks
number of ports, 4, Hose Connections: Front-left, Rear-left-right
Synonym(e):
Synthware M431004, vacuum manifold, vacuum unit
Größe auswählen
1 EA
€ 753,00
€ 753,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
€ 753,00
About This Item
UNSPSC-Code:
40161501
NACRES:
NB.51
€ 753,00
Check Cart for Availability
Leistungsmerkmale
number of ports 4
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware M431004
Allgemeine Beschreibung
With 4 mm double oblique, high vacuum hollow glass stopcocks precision ground with corresponding numbers on barrel and plug. Lower vacuum chambers on stopcock are evacuated with a separate hole in the plug in order to hold the plug in the stopcock barrels. Beams are 22 mm O.D. with front-left and rear-left-right 10 mm hose connections. Distance between stopcocks is 75 mm center to center.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)