Merck
SYNM181003
Synthware all-glass vacuum/inert gas manifold with hollow high vacuum stopcocks
number of ports, 3, Hose Connections: Front-left, Rear-right
Synonym(e):
Synthware M181003, vacuum manifold, vacuum unit
Größe auswählen
1 EA
€ 577,00
€ 577,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
€ 577,00
About This Item
UNSPSC-Code:
40161501
NACRES:
NB.51
€ 577,00
Check Cart for Availability
Leistungsmerkmale
number of ports 3
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware M181003
Allgemeine Beschreibung
With 4 mm double oblique, high vacuum hollow glass stopcocks precision ground with corresponding numbers on barrel and plug. Lower vacuum chambers on stopcock are evacuated with a separate hole in the plug in order to hold the plug in the stopcock barrels. Beams are 22 mmO.D. with front-left and rear-right 10 mm hose connections. Distance between stopcocks is 75 mm center to center.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)