Merck
SYNM141004
Synthware Vacuum/Inert Gas Manifold with double bank and high vacuum valves
number of ports, 4, Hose Connections: Front-right, Rear-left
Synonym(e):
Synthware M141004, vacuum manifold, vacuum unit
Größe auswählen
1 EA
€ 664,00
€ 664,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
€ 664,00
About This Item
UNSPSC-Code:
40161501
NACRES:
NB.51
€ 664,00
Check Cart for Availability
Leistungsmerkmale
number of ports 4
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware M141004
Allgemeine Beschreibung
General purpose double bank manifold for use with vacuum and inert gas. Supplied with 0-4 mm high vacuum valves for grease-free operation. Both banks are made from 22 mm medium wall tubing and have all banks open. Ports are spaced 100 mm center to center and have an O.D. of 10 mm. With front- right and rear- left 10 mm hose connections.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)