Merck
SLW3152/18A
Pyrex® Volumetric flask, heavy duty, class A, amber, USP/ISO/DIN tolerances,
capacity 500 mL
Synonym(e):
Pyrex, SciLabware 3152/18A
Größe auswählen
2 EA
€ 113,00
€ 113,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
2 EA
€ 113,00
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.43
€ 113,00
Check Cart for Availability
Materialien
borosilicate glass flask
Verbindung
joint ST/NS neck19/26
Leistungsmerkmale
accuracy class A (ISO 1042 and DIN 12664, United States Pharmacopoeia (USP),conformity ′H′ certified)
Verpackung
pkg of 2 ea
Hersteller/Markenname
SciLabware 3152/18
Gesamt H
260 mm
Kapazität
500 mL
Allgemeine Beschreibung
  • Complies with ISO 1042 and DIN 12664
  • Calibrated to the tolerance required by the United States Pharmacopoeia (USP) which are tighter than those of ISO/DIN standards
  • Manufactured from borosilicate glass with an amber coating on exterior surfaces to facilitate use with light sensitive solutions
  • Each flask is laser etched with an individual serial number, batch code and date of manufacturer
Leistungsmerkmale und Vorteile
  • Complies with ISO 1042 and DIN 12664
  • Manufactured from borosilicate glass 3.3
  • Calibrated to the tolerance required by the United States Pharmacopoeia (USP) which are tighter than those of ISO/DIN standards
  • With an amber coating on exterior surfaces to facilitate use with light sensitive solutions
  • Inscriptions in white enamel
  • Each flask is laser etched with an individual serial number batch code and date of manufacture
  • Batch certificate available to download online.
Rechtliche Hinweise
PYREX is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)