Größe auswählen
| Ihnen/SKU | Verfügbarkeit | Preis |
|---|---|---|
2 ea | Warenkorb auf Verfügbarkeit prüfen | € 120,00 |
Über diesen Artikel
€ 120,00
material
borosilicate glass 3.3 flask
joint
joint ST/NS neck19/26
description
calibrated to contain (TC, IN)
feature
class A DE-M, DIN EN ISO 1042. Calibrated to contain (TC, In).
packaging
pack of 2 ea
manufacturer/tradename
BRAND 37410
volume measuring range
500 mL, accuracy: 0.25 mL
capacity
500 mL
General description
Other Notes
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description volume 500 mL, accuracy: 0.25 mL | description volume 5 mL, accuracy: 0.04 mL | description volume 50 mL, accuracy: 0.06 mL | description volume 50 mL, accuracy: 0.08 mL |
| manufacturer/tradename BRAND 37410 | manufacturer/tradename BRAND 37401 | manufacturer/tradename BRAND 37405 | manufacturer/tradename BRAND 37406 |
| feature class A DE-M, DIN EN ISO 1042. Calibrated to contain (TC, In). | feature class A DE-M, DIN EN ISO 1042. Calibrated to contain (TC, In). | feature class A DE-M, DIN EN ISO 1042. Calibrated to contain (TC, In). | feature class A DE-M, DIN EN ISO 1042. Calibrated to contain (TC, In). |
| capacity 500 mL | capacity 5 mL | capacity 50 mL | capacity 50 mL |
| joint joint ST/NS neck19/26 | joint joint ST/NS neck10/19 | joint joint ST/NS neck12/21 | joint joint ST/NS neck14/23 |
| material borosilicate glass 3.3 flask | material borosilicate glass 3.3 flask, wide-neck | material borosilicate glass 3.3 flask | material borosilicate glass 3.3 flask, wide-neck |
| packaging pack of 2 ea | packaging pack of 2 ea | packaging pack of 2 ea | packaging pack of 2 ea |
Still not finding the right product?
Explore all of our products under BRAND® BLAUBRAND® Messkolben, PP-Stopfen, Braunglas
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?