Ansicht ändern
| Größe/SKU | Verfügbarkeit | Preis |
|---|---|---|
1 ea | Warenkorb auf Verfügbarkeit prüfen | € 372,00 |
Über diesen Artikel
NACRES:
NB.61
UNSPSC Code:
81141502
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltenproduct line
SynLectro®
size
3 mm × 20 mm × 60 mm
General description
SynLectro® Zinc Electrode is a 3x20x60 mm electrode designed for use with the SynLectro® Electrolysis Platform for synthetic electrochemistry.
SynLectro® Electrolysis Platform
The SynLectro® Electrolysis Platform is a portfolio of labware products and electrodes designed to simplify and standardize preparative scale electrosynthesis. Learn more about each component, designed with consistency, reproducibility, and safety in mind.
SynLectro® Electrolysis Platform
The SynLectro® Electrolysis Platform is a portfolio of labware products and electrodes designed to simplify and standardize preparative scale electrosynthesis. Learn more about each component, designed with consistency, reproducibility, and safety in mind.
Legal Information
SynLectro is a registered trademark of Merck KGaA, Darmstadt, Germany
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description size 3 mm × 20 mm × 60 mm | description size 3 mm × 20 mm × 60 mm | description size 3 mm × 20 mm × 60 mm | description size 3 mm × 20 mm × 60 mm |
| size 3 mm × 20 mm × 60 mm | size 3 mm × 20 mm × 60 mm | size 3 mm × 20 mm × 60 mm | size 3 mm × 20 mm × 60 mm |
| product line SynLectro® | product line SynLectro® | product line SynLectro® | product line SynLectro® |
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?