Merck
CLS9999918
Corning® disposable phenolic cap
18-415 thread
Synonym(e):
phenolic caps, threaded cap, threaded caps
Größe auswählen
1000 EA
€ 507,00
€ 507,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1000 EA
€ 507,00
About This Item
UNSPSC-Code:
41121500
NACRES:
NB.43
€ 507,00
Check Cart for Availability
Hersteller/Markenname
Corning 99999-18
Höhe
17.5 mm
Fitting
18-415 thread
Allgemeine Beschreibung
These disposable Corning caps are made from phenolic resin and are autoclavable at 121°C. Caps are provided with a white rubber liner.
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)