Merck
CLS7732CO
Pyrex® gum stability glass bombs
replacement cover for gum bomb, reusuable, non-sterile, 1/cs
Synonym(e):
lab equipment accessory
About This Item
UNSPSC-Code:
41121500
NACRES:
NB.43
Materialien
borosilicate glass (borosilicate glass code 7740)
Sterilität
non-sterile
Leistungsmerkmale
reusable
Verpackung
case of 1
pack of 1 ×
Hersteller/Markenname
Corning 7732-CO
AD × H
46 mm × 12 mm
Eignung
suitable for (the determination of gum stability of gasoline)
Allgemeine Beschreibung
Note: Replacement covers for gum bomb CLS7732. The cover prevents condensed vapors in bomb stem from contaminating the sample, but does not interfere with free flow of oxygen. Made in accordance with ASTM D-525.
These PYREX® bombs are for the determination of gum stability of gasoline. The cover prevents condensed vapors in bomb stem from contaminating the sample, but does not interfere with free flow of oxygen. Made in accordance with ASTM D-525.
Rechtliche Hinweise
PYREX is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)