Merck
CLS7077B1
Pyrex® Shorty serological pipettes, disposable, sterile, plugged, individually wrapped
volume 1.0 mL, accuracy: 0.01 mL
Synonym(e):
Corning® 7077B
Größe auswählen
500 EA
€ 382,00
€ 382,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
500 EA
€ 382,00
About This Item
UNSPSC-Code:
41121500
NACRES:
NB.43
€ 382,00
Check Cart for Availability
Materialien
borosilicate glass
Sterilität
sterile
Hersteller/Markenname
Corning 7077B-1
Messbereich Volumen
1.0 mL, accuracy: 0.01 mL
Länge
215 mm
Kapazität
1 mL
Allgemeine Beschreibung
These sterile disposable shorty Pyrex serological pipettes are calibrated "to deliver" (TD) with blow-out. They are ideal for use in confined areas such as hoods. The 5, 10 and 25 mL sizes have tooled tops and the 50 mL has a polypropylene top end. Negative graduations on each pipet make it applicable for both serological and measuring procedures.
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)