Merck
BR82121
BRAND® needle-valve stopcocks for burettes and automatic burettes with PTFE spindle
Lateral, for burette capacity 25 mL
Synonym(e):
Accessories, Burettes, Needle-valve stopcock, Stopcocks
Größe auswählen
1 EA
€ 46,30
€ 46,30
Voraussichtliches Versanddatum23. April 2025
Größe auswählen
Ansicht ändern
1 EA
€ 46,30
About This Item
UNSPSC-Code:
40141602
NACRES:
NB.43
€ 46,30
Voraussichtliches Versanddatum23. April 2025
Materialien
polystyrene
Leistungsmerkmale
for burette capacity 25 mL
Verpackung
pack of 1 ea
Hersteller/Markenname
BRAND 82121
Allgemeine Beschreibung
Interchangeable PTFE valve spindle, PP cap and retention device. Valve opening 0-25.5 mm.
Please note: PTFE valve spindle operates smoothly yet is tight, even without lubrication, so there is no grease residue on the burette. The valve spindle is replaceable. Tight seal without force, a gentle turn is all it takes to close the burette valve.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)