Merck
BR759500
BRAND® cuvette rack
Holds 16 cuvettes
Synonym(e):
Accessories, Cuvettes, Racks
Größe auswählen
1 EA
€ 12,20
€ 12,20
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
€ 12,20
About This Item
UNSPSC-Code:
41122800
NACRES:
NB.35
€ 12,20
Check Cart for Availability
Materialien
gray
polypropylene
Leistungsmerkmale
Holds 16 cuvettes
Verpackung
pack of 1 ea
Hersteller/Markenname
BRAND 759500
Größe
210 mm × 70 mm × 38 mm
Allgemeine Beschreibung
Our Grey PP Cuvette Rack is designed for efficient organization and storage of cuvettes in laboratory settings. Featuring numbered positions for easy identification, this rack measures 210 x 70 x 38 mm, making it compact yet functional. Constructed from durable polypropylene (PP), it is autoclavable at 121 °C, ensuring thorough sterilization for safe use. This cuvette rack is an essential tool for maintaining order and accessibility in your laboratory, enhancing workflow and productivity.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)