Merck
BR709109
BRAND® disposable BLAUBRAND® micropipettes, intraEND
volume 10 μL
Synonym(e):
Micropipettes, Pipettes, intraEND
About This Item
UNSPSC-Code:
41121500
NACRES:
NB.01
Leistungsmerkmale
DE-M marking, To Contain
Verpackung
pack of 1000 ea (4 boxes of 250)
pkg of (4 boxes of 250)
Hersteller/Markenname
BRAND 709109
Länge
32 mm
Kapazität
10 μL
Volumen
10 μL
Allgemeine Beschreibung
DE-M marking certified for volumes greater than 5 μl. Volume defined by end-to-end filling. Calibrated to contain (TC, In).
Rechtliche Hinweise
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)