Merck
BR704580
BRAND® discharge tube for QuikSip BT-Aspirator PP/EPDM
Synonym(e):
Accessories, Bottle-top Dispenser, Dispenser, QuikSip
About This Item
UNSPSC-Code:
41121500
NACRES:
NB.01
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Materialien
polypropylene
Verpackung
pack of 1 ea
Hersteller/Markenname
BRAND 704580
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 4
Dieser Artikel
ZRB1620ZRB1697ZRB1491
Gene Information
human ... IFNB1(3456)
Gene Information
-
Gene Information
-
Gene Information
human ... POLB(5423)
mouse ... Polb(18970)
rat ... Polb(29240)
biological source
rabbit
biological source
rabbit
biological source
rabbit
biological source
rabbit (recombinant)
species reactivity
human
species reactivity
human
species reactivity
rat, mouse, human
species reactivity
human, rat, mouse
antibody form
purified antibody
antibody form
purified antibody
antibody form
purified antibody
antibody form
purified antibody
clone
4F5, monoclonal, recombinant monoclonal
clone
recombinant monoclonal
clone
recombinant monoclonal
clone
3F21, monoclonal, recombinant monoclonal
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
QuikSip is a trademark of BRAND GMBH + CO KG
  • Hier finden Sie alle aktuellen Versionen:
    Analysenzertifikate (COA)